Learn More
S-Curve™ Four Compartment Glove Dispenser with Single Lid
Product Code. 16833250
Encompass
Change view
Click to view available options
Material:
Acrylic
Product Code. Material
16833250 Acrylic
1 options
Product Code. 16833250 Supplier S-Curve™ Supplier No. GD405
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Four compartment dispenser includes a single, sloping lid with a full-length hinge to maintain HEPA flow.
  • Single, sloping lid with full-length hinge to maintain HEPA flow
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 21 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 4
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.