missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fostamatinib, MedChemExpress
Description
Fostamatinib (R788) is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
Specifications
Specifications
| Chemical Name or Material | Fostamatinib |
| CAS | 901119-35-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H26FN6O9P |
| Quantity | 100 mg |
| Synonym | R788 |
| Solubility Information | DMSO : 62.5 mg/mL (107.67 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | COC1=CC(NC2=NC=C(C(NC3=NC(N4COP(O)(O)=O)=C(C=C3)OC(C)(C)C4=O)=N2)F)=CC(OC)=C1OC |
| Molecular Weight (g/mol) | 580.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?