missing translation for 'onlineSavingsMsg'
Learn More

Fosinopril sodium, MedChemExpress

Product Code. 16448276
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16448276

Brand: Medchem Express HYB0382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fosinopril Sodium is the ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor, used for the treatment of hypertension and some types of chronic heart failure.

Fosinopril Sodium is the ester prodrug of an angiotensin-converting enzyme (ACE) inhibitor, used for the treatment of hypertension and some types of chronic heart failure.

Specifications

Chemical Name or Material Fosinopril sodium
Physical Form Solid
CAS 88889-14-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C30H45NNaO7P
For Use With (Application) COVID-19-anti-virus
Synonym SQ28555
Solubility Information H2O : 33.33 mg/mL (56.91 mM; Need ultrasonic) ∣Ethanol : 16.67 mg/mL (28.46 mM; Need ultrasonic) ∣DMSO : 1.43 mg/mL (2.44 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@H]1N(C(C[P@@](CCCCC2=CC=CC=C2)(O[C@H](OC(CC)=O)C(C)C)=O)=O)C[C@H](C3CCCCC3)C1
Molecular Weight (g/mol) 585.64
Formula Weight 585.64
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.