missing translation for 'onlineSavingsMsg'
Learn More

Fosinopril Acyl-Beta-D-Glucuronide, TRC

Product Code. 30395804
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395804 10 mg 10mg
30390878 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395804 Supplier Toronto Research Chemicals Supplier No. TRCF72780510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fosinopril Acyl-beta-D-Glucuronide
Analyte or Component Names Fosinopril Acyl-beta-D-Glucuronide
Shipping Condition Room Temperature
Molecular Formula C36H54NO13P
Synonym (4S)-4-Cyclohexyl-1-[[(R)-[(1S)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetyl]-L-proline β-D-Glucopyranuronosyl Ester
InChI Formula InChI=1S/C36H54NO13P/c1-4-28(39)47-35(22(2)3)50-51(46,18-12-11-15-23-13-7-5-8-14-23)21-27(38)37-20-25(24-16-9-6-10-17-24)19-26(37)34(45)49-36-31(42)29(40)30(41)32(48-36)33(43)44/h5,7-8,13-14,22,24-26,29-32,35-36,40-42H,4,6,9-12,15-21H2,1-3H3,(H,43,44)/t25-,26+,29+,30+,31-,32+,35+,36+,51-/m1/s1
Solution Type Neat
SMILES O=[P@@](CC(N1[C@H](C(O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O2)=O)C[C@@H](C3CCCCC3)C1)=O)(O[C@@H](C(C)C)OC(CC)=O)CCCCC4=CC=CC=C4
IUPAC Name (2S,3S,4S,5R,6S)-6-(((2S,4S)-4-cyclohexyl-1-(2-((R)-((S)-2-methyl-1-(propionyloxy)propoxy)(4-phenylbutyl)phosphoryl)acetyl)pyrrolidine-2-carbonyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 739.79
Quantity 10 mg
Formula Weight 739.99
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.