missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin Phenylethylamine, TRC

Product Code. 30403313
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403313 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403313 Supplier Toronto Research Chemicals Supplier No. TRCF727510500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fosfomycin (R)-1-Phenethylamine Salt
CAS 25383-07-7
Molecular Formula C8 H11 N . C3 H7 O4 P
Quantity 500 mg
Synonym Phosphonic acid, P-[(2R,3S)-3-methyl-2-oxiranyl]-, compd. with (αR)-α-methylbenzenemethanamine (1:1), Phosphonic acid, (1,2-epoxypropyl)-, cis-(-)-, compd. with (+)-α-methylbenzylamine (1:1) (8CI), Phosphonic acid, (3-methyloxiranyl)-, (2R-cis)-, compd. w
InChI Formula InChI=1S/C8H11N.C3H7O4P/c1-7(9)8-5-3-2-4-6-8;1-2-3(7-2)8(4,5)6/h2-7H,9H2,1H3;2-3H,1H3,(H2,4,5,6)/t7-;2-,3+/m10/s1
SMILES C[C@@H](N)c1ccccc1.C[C@@H]2O[C@@H]2P(=O)(O)O
IUPAC Name [(2R,3S)-3-methyloxiran-2-yl]phosphonic acid;(1R)-1-phenylethanamine
Molecular Weight (g/mol) 259.239
Formula Weight 259.097
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.