missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin calcium, MedChemExpress

Codice prodotto. 16498586
Encompass_Preferred
Cambia vista
Click to view available options
Quantity:
100 mg
Dimensione della confezione:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
16498586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16498586 Fornitore Medchem Express N. del fornitore HYB1075

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Fosfomycin (MK-0955) calcium is a blood-brain barrier penetrating, broad-spectrum antibiotic by irreversibly inhibiting an early stage in cell wall synthesis. Fosfomycin calcium shows both in vivo and in vitro activity against a wide range of bacteria, including multidrug-resistant (MDR), extensively drug-resistant (XDR), and pan-drug-resistant (PDR) bacteria.

Fosfomycin (MK-0955) calcium is a blood-brain barrier penetrating, broad-spectrum antibiotic by irreversibly inhibiting an early stage in cell wall synthesis. Fosfomycin calcium shows both in vivo and in vitro activity against a wide range of bacteria, including multidrug-resistant (MDR), extensively drug-resistant (XDR), and pan-drug-resistant (PDR) bacteria.

Spécification

Chemical Name or Material Fosfomycin calcium
Physical Form Powder
CAS 26016-98-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H5CaO4P
For Use With (Application) COVID-19-immunoregulation
Synonym MK-​0955 calcium
Solubility Information H2O : 50 mg/mL (283.90 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES C[C@H]1[C@@H](P([O-])([O-])=O)O1.[Ca+2]
Molecular Weight (g/mol) 176.12
Formula Weight 176.12
Percent Purity 98.0%
Grade Research
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.