missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosfestrol Tetrasodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Fosfestrol Tetrasodium |
| CAS | 4719-75-9 |
| Molecular Formula | C18 H18 O8 P2 . 4 Na |
| Quantity | 10 mg |
| Synonym | Fosfestrol Tetrasodium Salt, Phenol, 4-[(1E)-1-ethyl-2-[4-(phosphonooxy)phenyl]-1-buten-1-yl]-, 1-(dihydrogen phosphate), sodium salt (1:4), 4,4'-Stilbenediol, α,α'-diethyl-, bis(dihydrogen phosphate), tetrasodium salt, (E)- (8CI), 4,4'-Stilbenediol, α,α'-diethyl-, diphosphate tetras |
| InChI Formula | InChI=1S/C18H22O8P2.4Na/c1-3-17(13-5-9-15(10-6-13)25-27(19,20)21)18(4-2)14-7-11-16(12-8-14)26-28(22,23)24;;;;/h5-12H,3-4H2,1-2H3,(H2,19,20,21)(H2,22,23,24);;;;/q;4*+1/p-4/b18-17+;;;; |
| SMILES | [Na+].[Na+].[Na+].[Na+].CC\C(=C(\CC)/c1ccc(OP(=O)([O-])[O-])cc1)\c2ccc(OP(=O)([O-])[O-])cc2 |
| IUPAC Name | tetrasodium;[4-[(E)-4-(4-phosphonatooxyphenyl)hex-3-en-3-yl]phenyl] phosphate |
| Molecular Weight (g/mol) | 516.237 |
| Formula Weight | 516.007 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?