missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FosB Monoclonal Antibody (83B1138)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA141121
This item is not returnable.
View return policy
Description
Suggested positive control: Hela whole cell extract, HeLa cell lysate.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
Specifications
| FosB | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.05% sodium azide | |
| P53539 | |
| Fosb | |
| Synthetic peptide corresponding to amino acids 2-15 of human FosB. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 83B1138 | |
| Unconjugated | |
| Fosb | |
| activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction