missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FosB/G0S3 Antibody (83B1138), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56530AF405
This item is not returnable.
View return policy
Description
FosB/G0S3 Monoclonal specifically detects FosB/G0S3 in Human samples. It is validated for Western Blot.
Specifications
| FosB/G0S3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| activator protein 1, AP-1, DKFZp686C0818, FBJ murine osteosarcoma viral oncogene homolog B, G0S3G0/G1 switch regulatory protein 3, GOS3, GOSB, MGC42291, oncogene FOS-B, protein fosB | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, a 46 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 83B1138 | |
| Western Blot | |
| P53539 | |
| FOSB | |
| This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB. | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 2354 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction