missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosamprenavir Calcium Salt, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Fosamprenavir Calcium Salt (GW433908G) is a phosphate ester prodrug of the antiretroviral protease inhibitor Amprenavir, with improved solubility. Anti-HIV infection.
Specifications
Specifications
| Chemical Name or Material | Fosamprenavir Calcium Salt |
| CAS | 226700-81-8 |
| Color | off White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H34CaN3O9PS |
| Quantity | 1 mg |
| Synonym | GW433908G |
| Solubility Information | DMSO : 50 mg/mL (80.17 mM; Need ultrasonic) ∣H2O : 0.25 mg/mL (0.40 mM; Need ultrasonic and warming) |
| SMILES | O=C(O[C@@H]1COCC1)N[C@@H](CC2=CC=CC=C2)[C@H](OP([O-])([O-])=O)CN(S(=O)(C3=CC=C(N)C=C3)=O)CC(C)C.[Ca+2] |
| Molecular Weight (g/mol) | 623.67 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction