Learn More
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe
Product Code. 11962129
Click to view available options
Volume (Metric):
25 mL
50 mL
Includes:
Capillary Tube
Straight Stopcock
Unit Size:
Each
Product Code. 11962129
Brand: Poulten & Graf GmbH 2 01246
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe are used to measure gas volume.
  • Clear neutral glass
  • Calibrated cylinders
  • Acid and alkali resistant amber stain diffused graduation
  • Amber graduation
Specifications
Autoclavable Autoclavable
Material Glass (Tip), Neutral Glass, Neutral Glass (Barrel), Neutral Glass (Plunger)
Color Clear
Includes Straight Stopcock
Product Type FORTUNA Precision Gas Syringe with straight stopcock
Disposable Reusable
Graduations Amber
Quantity 1
Volume (Metric) 50 mL
Graduated Yes
Sterility Non-sterile