missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser

Product Code. 11788286
Change view
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
6 to 30 mL
Includes:
PTFE-coated Piston, Stopcock and Ventilator Valve
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes Unit Size
11788286 10 to 50 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
10719212 2 to 10 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11778286 6 to 30 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
10341222 1 to 5 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11768286 0.4 to 2 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. Afficher la politique du retour.
Code produit 11788286 Fournisseur Poulten Graf™ Code fournisseur 101 10748S
Estimated Shipment: 11-01-2027
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.

For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.

  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With certificate, individually numbered
  • Glass dispensing cylinder is coated with plastic to provide breakage protection
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment

Spécification

Product Type Outfix Safety
Volume (Metric) 10 to 50 mL
Includes PTFE-coated Piston, Stopcock and Ventilator Valve
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.