Learn More
Poulten Graf™ Fortuna™ Glass 1 Mark Bulb Pipette
Description
- Short practical delivery time
- Tips and suction tubes reinforced
- Acid and alkali resistant
Specifications
Specifications
| Class | AS |
| Material | Glass |
| Graduated | Yes |
| Graduation Color | Blue |
| Quantity | 1 Pack |
| Volume (Metric) | 2 mL |
| Type | Bulb Pipet |