Learn More
Poulten Graf™ Fortuna™ Glass 1 Mark Bulb Pipette
Product Code. 10371222
Change view
Click to view available options
Volume (Metric):
2 mL
10 mL
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10371222 10 mL Each
15922110 2 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10371222 Supplier Poulten Graf™ Supplier No. 1 2303604
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Individually marked with the time of flow to transfer. Poulten Graf™ Fortuna™ Glass One Mark Bulb Pipettes are manufactured from clear soda lime glass and come with a batch certificate of conformity.
  • Short practical delivery time
  • Tips and suction tubes reinforced
  • Acid and alkali resistant
Specifications
Class AS
Material Glass
Graduated Yes
Graduation Color Amber
Quantity 1 Pack
Volume (Metric) 10 mL
Type Bulb Pipet