missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Forskolin

Product Code. p-4783468
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18782764

Brand: Tocris Bioscience™ 1099/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adenylyl cyclase activator

Forskolin, or coleonol, is a cell-permeable, potent, reversible and rapid activator of adenylyl cyclase, an enzyme that converts ATP to cAMP and pyrophosphate (EC50 = 0.5 μM). In combination with 5 other compounds, forskolin enables chemical reprogramming of mouse embryonic fibroblasts to pluripotent stem cells, without genetic factors: Valproic acid, sodium salt (Cat. No. 2815), RepSox (Cat. No. 3742), Tranylcypromine hydrochloride (Cat. No. 3852), CHIR 99021 (Cat. No. 4423) and 3-Deazaneplanocin A hydrochloride (Cat. No. 4703). In combination with IBMX (Cat. No. 2845), forskolin induces neural differentiation of mesenchymal stem cells by downregulating neuron restrictive silencer factor. In a rat model of Huntington's disease-like neurodegeneration, forskolin also exhibits neuroprotective effects. Hypotensive and vasodilatory agent.

Water-soluble analog, NKH 477 (Cat. No. 1603), and Forskolin synthesized to Ancillary Material Grade also available.

For more information about how Forskolin may be used, see our protocol: Transdifferentiating Fibroblasts into Neurons.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7

Specifications

Quantity 10 mg
Formula Weight 410.51
Purity >98%
Molecular Weight (g/mol) 410.507
Chemical Name or Material (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1H-naphtho[2,1-b]pyran-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.