missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forskolin (From Coleus Forskohlii), Fisher BioReagents
Description
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
Specifications
Specifications
| Melting Point | 228°C |
| Color | White |
| Packaging | Amber Glass |
| Quantity | 5 mg |
| Merck Index | 15, 2460 |
| Formula Weight | 410.5 |
| Physical Form | Solid |
| Chemical Name or Material | Forskolin, From Coleus forskohlii |
Safety and Handling
Product Identifier
- Forskolin
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?