missing translation for 'onlineSavingsMsg'
Learn More

Forskolin (From Coleus Forskohlii), Fisher BioReagents

Product Code. 11453074
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Packaging:
Amber Glass
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11453074 1 mg Amber Glass 1mg
11473074 5 mg Amber Glass 5mg
11463074 10 mg Amber Glass 10mg
10556394 25 mg Amber Glass 25mg
10615445 50 mg Amber Glass 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11453074 Supplier Fisher BioReagents Supplier No. BP25201
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Melting Point 228°C
Color White
Packaging Amber Glass
Quantity 1 mg
Merck Index 15, 2460
Formula Weight 410.5
Physical Form Solid
Chemical Name or Material Forskolin, From Coleus forskohlii
compliance-icons
Product Identifier
  • Forskolin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.