missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Forskolin

Product Code. 18734277
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18734277 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18734277 Supplier R&D Systems™ Supplier No. TB1099RMU/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Forskolin synthesized to Ancillary Material Grade

TB1099-RMU is an Ancillary Material Grade version of Forskolin (Cat. No. 1099). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 R,4 aR,5 S,6 S,6 AS,10 S,10 aR,10 bS)-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10 b-trihydroxy-3,4 A,7,7,10 A-pentamethyl-1 H-naphtho[2,1-b]pyran-1-one
CAS 66575-29-9
Quantity 10 mg
Product Line Tocris
Target Adenylyl Cyclase Activators
Molecular Formula C22H34O7
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.