missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Forskolin

Product Code. 17180022
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17180022 10 mg 10mg
17149971 25 mg 25mg
17170022 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17180022 Supplier STEMCELL Technologies Supplier No. 72114
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

cAMP pathway activator; Activates adenylyl cyclase

  • Forskolin is a cell-permeable diterpene that directly activates adenylyl cyclase (IC50 = 41 nM), the enzyme that produces cyclic adenosine monophosphate (cAMP), which as a result raises cAMP levels in the cell
  • cAMP is an important second messenger involved in many signal transduction pathways, including activation of protein kinase A (PKA; Awad et al.; Robbins et al.)

Specifications

CAS 66575-29-9
Synonym Coleonol; HL 362; L 75-1362B; NSC 357088; NSC 375489
Quantity 10 mg
Molecular Formula C22H34O7
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.