missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Forskolin

Product Code. p-7217354
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
This item is not returnable. View return policy

Product Code. 17149971

Brand: STEMCELL Technologies 1000249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cAMP pathway activator; Activates adenylyl cyclase

  • Forskolin is a cell-permeable diterpene that directly activates adenylyl cyclase (IC50 = 41 nM), the enzyme that produces cyclic adenosine monophosphate (cAMP), which as a result raises cAMP levels in the cell
  • cAMP is an important second messenger involved in many signal transduction pathways, including activation of protein kinase A (PKA; Awad et al.; Robbins et al.)

Specifications

CAS 66575-29-9
Synonym Coleonol; HL 362; L 75-1362B; NSC 357088; NSC 375489
Quantity 25 mg
Molecular Formula C22H34O7
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.