missing translation for 'onlineSavingsMsg'
Learn More

Foropafant, MedChemExpress

Product Code. 16465418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465418 10 mg 10mg
16453336 25 mg 25mg
16408137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465418 Supplier Medchem Express Supplier No. HY128694
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Foropafant (SR27417) highly potent, competitive, selective and orally active antagonist of platelet-activating factor (PAF) receptor, with a Ki value of 57 pM for [3H]PAF binding, at least 5-fold lower than that of unlabeled PAF itself. Foropafant potently inhibits PAF-induced aggregation of rabbit and human platelets.

Foropafant (SR27417) highly potent, competitive, selective and orally active antagonist of platelet-activating factor (PAF) receptor, with a Ki value of 57 pM for [3H]PAF binding, at least 5-fold lower than that of unlabeled PAF itself. Foropafant potently inhibits PAF-induced aggregation of rabbit and human platelets.

Specifications

Chemical Name or Material Foropafant
CAS 136468-36-5
Color White
Purity Grade Notes Research
Molecular Formula C28H40N4S
Quantity 10 mg
Synonym SR27417
Solubility Information DMSO : 100 mg/mL (215.18 mM; Need ultrasonic)
SMILES CC(C)C1=CC(C(C)C)=C(C2=CSC(=N2)N(CCN(C)C)CC2=CC=CN=C2)C(=C1)C(C)C
Molecular Weight (g/mol) 464.72
Formula Weight 464.72
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.