missing translation for 'onlineSavingsMsg'
Learn More

Forodesine hydrochloride, MedChemExpress

Product Code. 16469547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16469547

Brand: Medchem Express HY16209

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Forodesine hydrochloride (BCX-1777 hydrochloride) is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor with IC50 values ranging from 0.48 to 1.57 nM for human, mouse, rat, monkey and dog PNP. Forodesine hydrochloride is a potent human lymphocyte proliferation inhibitor. Forodesine hydrochloride could induce apoptosis in leukemic cells by increasing the dGTP levels.

Forodesine hydrochloride (BCX-1777 hydrochloride) is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor with IC50 values ranging from 0.48 to 1.57 nM for human, mouse, rat, monkey and dog PNP. Forodesine hydrochloride is a potent human lymphocyte proliferation inhibitor. Forodesine hydrochloride could induce apoptosis in leukemic cells by increasing the dGTP levels.

Specifications

Chemical Name or Material Forodesine hydrochloride
CAS 284490-13-7
Color Brown
Purity Grade Notes Research
Molecular Formula C11H15ClN4O4
Quantity 5 mg
Synonym BCX-1777 hydrochlorideImmucillin-H hydrochloride
Solubility Information H2O : 100 mg/mL (330.35 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (33.03 mM; Need ultrasonic)
SMILES O=C1C(NC=C2[C@@H]3N[C@H](CO)[C@@H](O)[C@H]3O)=C2NC=N1.Cl
Molecular Weight (g/mol) 302.71
Formula Weight 302.71
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.