missing translation for 'onlineSavingsMsg'
Learn More

(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water

Product Code. 11419223
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419223 10 g 10g
11488900 25 g 25g
11498900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419223 Supplier Thermo Scientific Alfa Aesar Supplier No. A11709.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2136-75-6 | C20H17OP | 304.329 g/mol

(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2136-75-6
Molecular Formula C20H17OP
Molecular Weight (g/mol) 304.329
MDL Number MFCD00006994
InChI Key CQCAYWAIRTVXIY-UHFFFAOYSA-N
Synonym 2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l
PubChem CID 75051
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
SMILES C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 183°C to 188°C
Quantity 10 g
Beilstein 523797
Sensitivity Air sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 304.33
Percent Purity 97%
Concentration or Composition (by Analyte or Components) May contain up to ≈3% water
Chemical Name or Material (Formylmethylene)triphenylphosphorane
compliance-icons
Product Identifier
  • (Formylmethylene)triphenylphosphorane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: (Triphenylphosphoranylidene)acetaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.