missing translation for 'onlineSavingsMsg'
Learn More

Formylglycinamide Ribotide (Technical Grade), TRC

Product Code. 30399926
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399926 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399926 Supplier Toronto Research Chemicals Supplier No. TRCF7004425MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formylglycinamide Ribotide
CAS 349-34-8
Molecular Formula C8 H15 N2 O9 P
Quantity 5 mg
Synonym 2-(Formylamino)-N-(5-O-phosphono-β-D-ribofuranosyl)acetamide, 2-Formamido-N-β-D-ribofuranosylacetamide 5'-(Dihydrogen phosphate), 5'-Phosphoribosyl-N-formylglycinamide, FGAR, Formylglycinamide Ribonucleotide, N-Formylglycinamide Ribonucleotide, Phosphoribosylformylglycinamide, α-N-Formylglycinamide Ribonucleotide
InChI Formula InChI=1S/C8H15N2O9P/c11-3-9-1-5(12)10-8-7(14)6(13)4(19-8)2-18-20(15,16)17/h3-4,6-8,13-14H,1-2H2,(H,9,11)(H,10,12)(H2,15,16,17)/t4-,6-,7-,8?/m1/s1
SMILES O[C@H]1[C@@H](O)C(NC(=O)CNC=O)O[C@@H]1COP(=O)(O)O
IUPAC Name [(2R,3S,4R)-5-[(2-formamidoacetyl)amino]-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
Molecular Weight (g/mol) 314.187
Formula Weight 314.052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.