missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formylglycinamide Ribotide (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Formylglycinamide Ribotide |
| CAS | 349-34-8 |
| Molecular Formula | C8 H15 N2 O9 P |
| Quantity | 0.5 mg |
| Synonym | 2-(Formylamino)-N-(5-O-phosphono-β-D-ribofuranosyl)acetamide, 2-Formamido-N-β-D-ribofuranosylacetamide 5'-(Dihydrogen phosphate), 5'-Phosphoribosyl-N-formylglycinamide, FGAR, Formylglycinamide Ribonucleotide, N-Formylglycinamide Ribonucleotide, Phosphoribosylformylglycinamide, α-N-Formylglycinamide Ribonucleotide |
| InChI Formula | InChI=1S/C8H15N2O9P/c11-3-9-1-5(12)10-8-7(14)6(13)4(19-8)2-18-20(15,16)17/h3-4,6-8,13-14H,1-2H2,(H,9,11)(H,10,12)(H2,15,16,17)/t4-,6-,7-,8?/m1/s1 |
| SMILES | O[C@H]1[C@@H](O)C(NC(=O)CNC=O)O[C@@H]1COP(=O)(O)O |
| IUPAC Name | [(2R,3S,4R)-5-[(2-formamidoacetyl)amino]-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| Molecular Weight (g/mol) | 314.187 |
| Formula Weight | 314.052 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?