missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Formyl-Histone H1.4 (Lys63) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17792804
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17792804 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17792804 Supplier Invitrogen™ Supplier No. PA5121411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Formyl-Histone H1.4 (Lys63)
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene H1-4
Gene Accession No. P10412
Gene Alias dJ221C16.5; H1 histone family, member 4; H1 VAR.2; H1.12 linker histone, cluster member; H1.4; H1-12; H14; H1-4; H1d; H1e; H1f4; H1s-4; H1var2; Hist1h1c; Hist1h1d; Hist1h1e; histone 1, H1e; Histone 1d; Histone cluster 1; histone cluster 1, H1d; histone cluster 1, H1e; histone H1.2; histone H1.4; Histone H1b; histone H1e; histone H1s-4; MGC116819
Gene Symbols H1-4, HIST1H1E
Host Species Rabbit
Immunogen Synthesized peptide derived from Human Histone H1.4 protein (57-69aa).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3008
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.