missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid Solution 0.25M, Certified, for HPLC, Fisher Chemical

Product Code. 10000112 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10000112 500 mL Plastic Coated Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10000112 Supplier Fisher Chemical Supplier No. J/4200/PB08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 64-18-6
Color Colorless
Packaging Plastic Coated Glass bottle
Molecular Formula CH2O2
MDL Number 3297
Quantity 500 mL
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES C(=O)O
IUPAC Name formic acid
Molecular Weight (g/mol) 46.025
PubChem CID 284
ChEBI CHEBI:30751
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.