missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Reagent grade, Honeywell

Product Code. 15697850 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15697850 500 mL 500mL
15677850 100 mL 100mL
15687850 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15697850 Supplier Honeywell Research Chemicals Supplier No. F0507500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Formula Weight 46.03g/mol
Percent Purity ≥95%
Grade Reagent
Density 1.22g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.57
Flash Point 49°C (120.2°F)
Ignition Point 1004°F
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8 mmHg (20°C)
Chemical Name or Material Formic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.