missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10063427 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
0.5 mL
1 mL
10 Ampules
2 mL
50 mL
Packaging:
Ampule
Ampules
Nalgene™ Poly Bottle
Unit Size:
0.5mL
10 ampoules
1mL
2mL
50mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10063427 2 mL Ampule 2mL
10780320 0.5 mL Ampule 0.5mL
10473038 1 mL Ampules 1mL
10797488 10 Ampules Ampules 10 ampoules
10596814 50 mL Nalgene™ Poly Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10063427 Supplier Fisher Chemical Supplier No. A1172AMP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable as a mobile phase additive for LC/MS and UHPLC/MS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid

Specifications

Absorbance 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Assay Percent Range ≥99.0 %
Linear Formula HCOOH
Merck Index 15, 4269
SMILES C(=O)O
Molecular Weight (g/mol) 46.025
Formula Weight 46.02
Percent Purity ≥99.0%
Grade OPTIMA™ LC-MS
Density 1.220g/cm³
Quantity 2 mL
Packaging Ampule
Physical Form Liquid
Residue after Evaporation 1ppm max.
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic Acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid, OPTIMA LC/MS Grade (99.5%)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.