missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, LC/MS grade, Thermo Scientific

Product Code. p-200109918 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
2 mL
5 mL
50 mL
10 x 1 mL
Unit Size:
1mL
2mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 30280953

Brand: Thermo Scientific T001170001

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Item Name Formic Acid
Form Liquid
Reagent Type Ion Pairing Reagent
Quantity 1 mL
Formulation 99% Formic Acid
For Use With LC/MS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.