missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, LC/MS grade, Thermo Scientific
GREENER_CHOICE

Product Code. 30280953 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
5 mL
50 mL
10 x 1 mL
Unit Size:
1mL
2mL
50mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30280953 1 mL 1mL
30280969 2 mL 2mL
30280955 5 mL 5mL
30280925 50 mL 50mL
30280965 10 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30280953 Supplier Thermo Scientific Supplier No. T001170001
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Item Name Formic Acid
Form Liquid
Reagent Type Ion Pairing Reagent
Quantity 1 mL
Formulation 99% Formic Acid
For Use With LC/MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.