missing translation for 'onlineSavingsMsg'
Learn More

Formic acid hydrazide, 90%

Product Code. 10305872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10305872 100 g Glass bottle 100g
10468092 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10305872 Supplier Thermo Scientific Acros Supplier No. 376061000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-84-0 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-84-0
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
MDL Number MFCD00007589
InChI Key XZBIXDPGRMLSTC-UHFFFAOYSA-N
Synonym formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
PubChem CID 12229
IUPAC Name formohydrazide
SMILES C(=O)NN

Specifications

Melting Point 51°C to 58°C
Color White
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula HCONHNH2
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 60.06
Percent Purity 90%
Physical Form Shiny Crystalline Flakes
Chemical Name or Material Formic acid hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • Formic acid hydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.