missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid-d2 (95% w/w in D2O), TRC

Product Code. 30404885
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Product Code. Quantity
30404885 5 g
30402989 10 g
30392739 1 g
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30404885

Brand: Toronto Research Chemicals TRCF6929035G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-42-3
InChI Formula InChI=1S/CH2O2/c2-1-3/h1H,(H,2,3)/i1D/hD
Molecular Formula CD2O2
Molecular Weight (g/mol) 48.04
SMILES [2H]OC(=O)[2H]
Synonym Formic Acid-d2, Perdeuterioformic Acid-d2
Concentration or Composition (by Analyte or Components) 0.95
Chemical Name or Material Formic Acid-d2

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.