missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid (Aldehyde-Free/Sequencing), Fisher BioReagents™

Product Code. 10736834
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10736834 500 mL Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10736834 Supplier Fisher BioReagents Supplier No. BP1215500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Formic Acid is used in Maxam-Gilbert DNA sequencing reactions and other biological applications.
TRUSTED_SUSTAINABILITY

Specifications

pH 2.1
CAS 64-18-6, 7732-18-5
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Assay Percent Range ≥88.0 %
Molecular Formula CH2O2
Linear Formula HCOOH
UN Number 1779
Merck Index 15, 4269
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES C(=O)O
IUPAC Name formic acid
Molecular Weight (g/mol) 46.025
PubChem CID 284
ChEBI CHEBI:30751
Formula Weight 46.02
Percent Purity ≥88.0%
Grade Sequencing
Density 1.220g/cm³
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic Acid, Aldehyde-free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid (> 85%)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.