missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, ACS Reagent, 88 to 91%, Honeywell™ Fluka™

Product Code. 15695380 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Poly Bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15695380 100 mL Poly Bottle 100mL
15645390 500 mL Poly Bottle 500mL
15615390 2.5 L Poly Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15695380 Supplier Honeywell-Fluka Supplier No. 399388100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent, 88-91%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

pH Acidic
Melting Point 4°C
Boiling Point ∼100.4°C
Color Colorless
Appearance Liquid
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Solubility Information Water soluble
Formula Weight 46.03g/mol
Percent Purity 88 to 91%
Density 1.22g/mL at 25°C
Quantity 100 mL
Explosion Limit 12% to 38%
Flash Point 48°C (118.4°F)
Ignition Point 1004°F
Odor Stinging
Packaging Poly Bottle
Refractive Index n20/D 1.370
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8mmHg (20°C)
Chemical Name or Material Formic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.