missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid in Acetonitrile, 0.1%, BAKER ANALYZED LC/MS, J.T. Baker™

Product Code. 13245503 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13245503 1 L Glass 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13245503 Supplier JT Baker Supplier No. 9824.1000GL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For more routine applications, this LC/MS solvent blend is function-tested and optimized for minimal impurities and interference-free baselines, giving trusted performance in the mobile phase, every time.

High purity solvents/reagents give optimal performance with minimal risk of contaminants that can limit accuracy and productivity, and they maximize the sensitivity and detecting power of instrumentation.

  • Application-optimized and function tested to enhance separation power and reproducibility
  • For use in more routine LC/MS applications
  • Tested for ESI+, UV-VIS absorbance, trace metals, residue after evaporation, assay and overall LC/MS suitability

Specifications

Absorbance Max. 0.1 at 240nm, Max. 0.01 at 260nm
Melting Point 0°C
Boiling Point 81°C
CAS 75-05-8, 64-18-6
Assay Percent Range 0.095 - 0.105%
Molecular Formula CH2O2
Linear Formula HCOOH
Synonym Methanoic acid, Carbonous Acid, Oxomethanol
Solubility Information Fully miscible in water
Formula Weight 46.03g/mol
Percent Purity 0.1%
Grade BAKER ANALYZED™ LC-MS
Density 0.9g/cm3
Quantity 1 L
Explosion Limit 4.4 - 16.0 Vol %
Flash Point 5°C
Ignition Point 525°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Assay v/v
Chemical Name or Material Formic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.