missing translation for 'onlineSavingsMsg'
Learn More

Formic acid ≥99%, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Product Code. p-7220853 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
2mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508664 2 mL 2mL
10103742 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15508664 Supplier JT Baker Supplier No. 9820.0002AM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tested for UHPLC-MS

  • LC-Gradient with Diode Array Detection (AU), 0.1% (v/v) aqueous solution:
    • at 220 nm max. 0.002
    • at 254 nm max. 0.002
  • LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), 0.1% (v/v) aqueous solution:
    • Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ppb
  • Ultraviolet Absorbance, 0.1% (v/v) aqueous solution (1.00-path vs water):
    • at 240 nm max. 0.1
    • at 260 nm max. 0.01

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic acid 100%
IUPAC Name formic acid
SMILES OC=O

Specifications

Assay Percent Range min. 99%
UN Number 1779
Quantity 2 mL
Flash Point 69°C
Residue after Evaporation max. 20 ppm
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.