missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 85%, TRC

Product Code. 30401962
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401962 10 g -
30399931 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401962 Supplier Toronto Research Chemicals Supplier No. TRCF69290010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula C H2 O2
Molecular Weight (g/mol) 46.03
Synonym Formic acid, Add-F, Add-F (silage additive), Amasil, Amasil 85, Ameisensaure, Aminic acid, Bilorin, Collo-Bueglatt, Collo-Didax, Ensilox, Formira, Formisoton, Formylic acid, Hydrogen carboxylic acid, Methanoic acid, Methanoic acid monomer, Myrmicyl, Sybest, Wonderbond Hardener M 600L
IUPAC Name formic acid
SMILES OC=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/CH2O2/c2-1-3/h1H,(H,2,3)
Formula Weight 46.0055
Chemical Name or Material Formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.