missing translation for 'onlineSavingsMsg'
Learn More

Formamidinesulfinic acid, 99%

Product Code. 10172910 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 10172910

Brand: Thermo Scientific Acros 119725000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1758-73-2 | CH4N2O2S | 108.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-73-2
Molecular Formula CH4N2O2S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00002397
InChI Key FYOWZTWVYZOZSI-UHFFFAOYSA-N
Synonym thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
PubChem CID 61274
IUPAC Name amino(imino)methanesulfinic acid
SMILES C(=N)(N)S(=O)O

Specifications

Melting Point 126.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NC(=NH)SO2H
Quantity 500 g
Beilstein 03, I, 36
Fieser 04,506; 05,668; 06,586
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents
Formula Weight 108.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Formamidinesulfinic acid, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formamidinesulfinic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.