missing translation for 'onlineSavingsMsg'
Learn More

Formamidine hydrochloride, 98%

Product Code. 15467448
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467448 5 g 5g
15477448 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467448 Supplier Thermo Scientific Alfa Aesar Supplier No. H60947.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6313-33-3 | CH4ClN2 | 79.51 g/mol

5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

Solubility
Soluble in Ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6313-33-3
Molecular Formula CH4ClN2
Molecular Weight (g/mol) 79.51
MDL Number MFCD00012865
InChI Key OEJWQRAWUVWBIE-UHFFFAOYSA-N
Synonym formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
PubChem CID 10313058
IUPAC Name methanimidamide;hydrochloride
SMILES [Cl].NC=N

Specifications

Melting Point 84°C to 87°C
Linear Formula HN=CHNH2·HCl
Quantity 5 g
Beilstein 3906935
Sensitivity Hygroscopic
Solubility Information Soluble in Ethanol.
Formula Weight 80.52
Percent Purity 98%
Chemical Name or Material Formamidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.