missing translation for 'onlineSavingsMsg'
Learn More

Formamide, TRC

Product Code. 30136568
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136568 250 mL 250mL
18062140 50 mL 50mL
18081640 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136568 Supplier Toronto Research Chemicals Supplier No. TRCF691480250ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula C H3 N O
Molecular Weight (g/mol) 45.04
Synonym Formamide, Carbamaldehyde, Formimidic acid, Methanamide, NSC 748
IUPAC Name formamide
SMILES NC=O

Specifications

Quantity 250 mL
InChI Formula InChI=1S/CH3NO/c2-1-3/h1H,(H2,2,3)
Formula Weight 45.0215
Percent Purity >95%
Chemical Name or Material Formamide
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.