missing translation for 'onlineSavingsMsg'
Learn More

Formamide, MedChemExpress

Product Code. 16499416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499416 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499416 Supplier Medchem Express Supplier No. HYY0842
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Formamide is an amide derived from formic acid and has been used as solvent for many ionic compounds.

Formamide is an amide derived from formic acid and has been used as solvent for many ionic compounds.

Specifications

Chemical Name or Material Formamide
CAS 75-12-7
Color Colorless
Quantity 100 g
Physical Form Liquid
Purity Grade Notes Research
Molecular Formula CH3NO
Synonym Methanamide Formimidic acid
Solubility Information H2O : 100 mg/mL (2220.25 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (2220.25 mM; Need ultrasonic)
SMILES O=CN
Molecular Weight (g/mol) 45.04
Formula Weight 45.04
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.