missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99%

Product Code. 11468933
Change view
Click to view available options
Quantity:
500 g
2500 g
10 kg
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468933 2500 g 2500g
11458933 500 g 500g
11478933 10 kg 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11468933 Supplier Thermo Scientific Alfa Aesar Supplier No. A11076.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coatings and in some personal care products.

Solubility
It is miscible with water.

Notes
Incompatible with bases, oxidizing agents, hydrogen peroxide, iodine, pyridine and sulphur oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Density 1.134
Quantity 2500 g
Flash Point 150°C (302°F)
Odor Ammonia-like
Refractive Index 1.4475
Assay Percent Range 99%
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Boiling Point 210°C
Solubility Information It is miscible with water.
Formula Weight 45.04
Percent Purity 99%
Chemical Name or Material Formamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.