missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Spectrophotometric Grade, ≥99%, Solstice

Product Code. 15682960
Change view
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15682960 2 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15682960 Supplier Honeywell-Riedel-de Haen Supplier No. 2958761L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 1°C to 3°C
CAS 75-12-7
Density 1.134g/mL at 25°C
Quantity 2 L
Flash Point 175°C (347°F)
Ignition Point 932°F
Packaging Glass Bottle
Refractive Index 1.447
Molecular Formula CH3NO
Linear Formula HCONH2
Vapor Pressure 0.08mmHg (20°C), 30mmHg (129°C)
MDL Number MFCD00007941
UN Number NONH for all modes of transport
Beilstein 505995
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
Boiling Point 210°C
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILES NC=O
IUPAC Name formamide
Molecular Weight (g/mol) 45.04
PubChem CID 713
ChEBI CHEBI:48431
Formula Weight 45.04g/mol
Percent Purity ≥99%
Grade Spectrophotometry
Chemical Name or Material Formamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.