missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99.6%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10617401
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Density 1.56 (Air = 1.0)
Quantity 1 kg
Infrared Spectrum Authentic
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Assay Percent Range 99.6%
Linear Formula HCONH2
Vapor Pressure Vapor Pressure: 0.08mbar at 20°C
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Formula Weight 45.04
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material Formamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'