missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99.5+%, extra pure

Product Code. 10427950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10427950 25 L Plastic Drum 25 L
10052370 1 L Glass Bottle 1 L
10592971 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.
Product Code. 10427950 Supplier Thermo Scientific Acros Supplier No. 181090250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Color Colorless to Yellow
Density 1.1330g/mL
Quantity 25 L
Flash Point 175°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4460 to 1.4480
Residue after Evaporation 0.05% max.
Assay Percent Range 99.5+%
Specific Gravity 1.133
Linear Formula HCONH2
Viscosity 3.75 mPa.s (20°C)
Water 0.1% max. (Coulometric)
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Boiling Point 210.0°C
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Percent Purity ≥99.5%
Grade Extra Pure
Chemical Name or Material Formamide, 99.5+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.