missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formalin-d2 (20% w/w in H2O), CDN
Click to view available options
Quantity:
10 g
20 g
Unit Size:
10g
20g
Specifications
Specifications
| Chemical Name or Material | Formaldehyde-d2 |
| CAS | 1664-98-8 |
| Solvent or Matrix | H2O |
| Molecular Formula | C 2H2 O |
| Quantity | 20 g |
| Synonym | Deuterated formaldehyde, Formalin-d2 |
| InChI Formula | InChI=1 S/CH2O/c1-2/h1H2/i1D2 |
| SMILES | [2 H]C(=O)[2 H] |
| IUPAC Name | dideuteriomethanone |
| Molecular Weight (g/mol) | 32.04 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction