missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP, 35 wt. %, Honeywell™ Riedel-de Haen™
BIOPHARMA

Catalog Number 15651470
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
1000 kg
Packaging:
Poly Bottle
Returnable Composite IBC
50-00-0
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
712
CHEBI:16842
C=O
50-00-0
CH2O
30.03
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
712
CHEBI:16842
C=O

Meets analytical specification of BP, Ph. Eur., 35 wt. %, contains 10% methanol as stabilizer

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Melting Point <-15°C
Color Colorless
Density 1.09g/mL at 25°C
pH 3.5 to 4.5 (at 20°C)
Boiling Point 93°C to 96°C
Flash Point 64°C (147.2°F)
Ignition Residue ≤0.01% (as SO4)
Packaging Poly Bottle
Linear Formula HCHO
Quantity 4 x 2.5 L
UN Number UN2209
Beilstein 1209228
IUPAC Name formaldehyde
Formula Weight 30.03g/mol
Percent Purity 35 wt. %
Chemical Name or Material Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'delimitedSupplementalStatement'
  • MIXTURE LIST-Contains : formaldehyde, methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'