missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP, 35 wt. %, Honeywell™ Riedel-de Haen™

Product Code. 15621470
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
1000 kg
Packaging:
Poly Bottle
Returnable Composite IBC
Unit Size:
1000kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15621470

Brand: Honeywell-Riedel-de Haen 155131000KG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., 35 wt. %, contains 10% methanol as stabilizer

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
SMILES C=O

Specifications

Melting Point <-15°C
Color Colorless
Density 1.09g/mL at 25°C
pH 3.5 to 4.5 (at 20°C)
Boiling Point 93°C to 96°C
Flash Point 64°C (147.2°F)
Ignition Residue ≤0.01% (as SO4)
Packaging Returnable Composite IBC
Linear Formula HCHO
Quantity 1000 kg
UN Number UN2209
Beilstein 1209228
IUPAC Name formaldehyde
Formula Weight 30.03g/mol
Percent Purity 35 wt. %
Chemical Name or Material Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.