missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, Honeywell
Click to view available options
Quantity:
1 L
2.5 L
60 kg
Specifications
Density | 1.09g/mL at 25°C |
Flash Point | 64°C (147.2°F) |
Linear Formula | HCHO |
Quantity | 2.5 L |
UN Number | 2209 |
Beilstein | 1209228 |
Formula Weight | 30.03g/mol |
Chemical Name or Material | Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP |
Safety and Handling
Product Identifier
- Formaldehyde solution
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : formaldehyde, methanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'